Quartz wool, coarse, 9-30 micron
Thermo Scientific Chemicals

Quartz wool, coarse, 9-30 micron

Filter aid
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Catalog number 393610100
Price (USD)
45.50
Each
Quantity:
10 g
Request bulk or custom format
Price (USD)
45.50
Each
Chemical Identifiers
CAS14808-60-7
IUPAC Namesilanedione
Molecular FormulaO2Si
InChI KeyVYPSYNLAJGMNEJ-UHFFFAOYSA-N
SMILESO=[Si]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Coarse fibers
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only