Alcohol 2-bromo-alfa-metilbencílico, 99 %, Thermo Scientific Chemicals
Alcohol 2-bromo-alfa-metilbencílico, 99 %, Thermo Scientific Chemicals
Alcohol 2-bromo-alfa-metilbencílico, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Alcohol 2-bromo-alfa-metilbencílico, 99 %, Thermo Scientific Chemicals

CAS: 5411-56-3 | C8H9BrO | 201.06 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
5 g
25 g
Envase:
Frasco de vidrio
Número de catálogo 393640050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS5411-56-3
IUPAC Name1-(2-bromophenyl)ethan-1-ol
Molecular FormulaC8H9BrO
InChI KeyDZLZSFZSPIUINR-UHFFFAOYNA-N
SMILESCC(O)C1=CC=CC=C1Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Liquid
Refractive index1.5590 to 1.5610 (20°C, 589 nm)
Appearance (Color)Clear yellow
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only