1-Butanol, for HPLC
1-Butanol, for HPLC
1-Butanol, for HPLC
Thermo Scientific Chemicals

1-Butanol, for HPLC

CAS: 71-36-3 | C4H10O | 74.123 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 393750025
Price (USD)
135.65
Special offer
Online exclusive
Ends: 30-Jun-2026
159.00
Save 23.35 (15%)
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
135.65
Special offer
Online exclusive
Ends: 30-Jun-2026
159.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS71-36-3
IUPAC Namebutan-1-ol
Molecular FormulaC4H10O
InChI KeyLRHPLDYGYMQRHN-UHFFFAOYSA-N
SMILESCCCCO
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.0003 %
Color scale=<10 APHA
Acidity=<0.0002 meq/g
UVat 210 nm A: =<0.699
UVat 245 nm A: =<0.046
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only