2-Trifluoromethoxyphenylboronic acid, 97%
2-Trifluoromethoxyphenylboronic acid, 97%
2-Trifluoromethoxyphenylboronic acid, 97%
Thermo Scientific Chemicals

2-Trifluoromethoxyphenylboronic acid, 97%

CAS: 175676-65-0 | C7H6BF3O3 | 205.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 393810050
Price (USD)
191.00
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
191.00
Each
Chemical Identifiers
CAS175676-65-0
IUPAC Name[2-(trifluoromethoxy)phenyl]boronic acid
Molecular FormulaC7H6BF3O3
InChI KeyAIJCNTOYZPKURP-UHFFFAOYSA-N
SMILESOB(O)C1=CC=CC=C1OC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)White to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only