5-Bromopicolinic acid, 97%
5-Bromopicolinic acid, 97%
5-Bromopicolinic acid, 97%
Thermo Scientific Chemicals

5-Bromopicolinic acid, 97%

CAS: 30766-11-1 | C6H3BrNO2 | 201.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 393990250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS30766-11-1
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only