gamma-Hexalactone, 98%
gamma-Hexalactone, 98%
gamma-Hexalactone, 98%
Thermo Scientific Chemicals

gamma-Hexalactone, 98%

CAS: 695-06-7 | C6H10O2 | 114.14 g/mol
数量:
25 g
100 g
500 g
製品番号(カタログ番号) 394171000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS695-06-7
Appearance (Color)Clear colorless to light yellow
Refractive index1.4380 to 1.4400 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only