Thermo Scientific Chemicals

Perfluoro-compound FC-770(TM), Thermo Scientific Chemicals

Catalog number: 394890250
25 mL, Each
Thermo Scientific Chemicals

Perfluoro-compound FC-770(TM), Thermo Scientific Chemicals

Catalog number: 394890250
25 mL, Each
Quantity

Chemical Identifiers

CAS
1093615-61-2
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Boiling point
90°C to 98°C (760 mmHg)
Density
1.76 to 1.80 g/ml (25°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text