Allylboronic acid pinacol ester, 98%
Allylboronic acid pinacol ester, 98%
Allylboronic acid pinacol ester, 98%
Thermo Scientific Chemicals

Allylboronic acid pinacol ester, 98%

CAS: 72824-04-5 | C9H17BO2 | 168.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 395130010
Price (EUR)
43,30
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
43,30
Each
Chemical Identifiers
CAS72824-04-5
IUPAC Name4,4,5,5-tetramethyl-2-(prop-2-en-1-yl)-1,3,2-dioxaborolane
Molecular FormulaC9H17BO2
InChI KeyYMHIEPNFCBNQQU-UHFFFAOYSA-N
SMILESCC1(C)OB(CC=C)OC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear colorless
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only