Raney™-Nickel, Activated Catalyst, 50% slurry in water
Raney™-Nickel, Activated Catalyst, 50% slurry in water
Raney™-Nickel, Activated Catalyst, 50% slurry in water
Thermo Scientific Chemicals

Raney™-Nickel, Activated Catalyst, 50% slurry in water

CAS: 7440-02-0 | Ni | 58.69 g/mol
製品番号(カタログ番号) 395921000
価格(JPY)
-
数量:
100 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
さらに表示
Aluminium (Al)=<8 %
pH8.5 to 12.0 (of supernatant)
Appearance (Form)Slurry in water
Nickel (Ni)>=92 %
Appearance (Color)Dark grey to black
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only