Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized
Thermo Scientific Chemicals

Cyclopentyl methyl ether, 99.9+%, extra pure, stabilized

CAS: 5614-37-9 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number 396132500
Price (USD)
49.65
Special offer
Online exclusive
Ends: 28-Dec-2026
58.00
Save 8.35
Each
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
49.65
Special offer
Online exclusive
Ends: 28-Dec-2026
58.00
Save 8.35
Each
Chemical Identifiers
CAS5614-37-9
IUPAC Namemethoxycyclopentane
Molecular FormulaC6H12O
InChI KeySKTCDJAMAYNROS-UHFFFAOYSA-N
SMILESCOC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.4190 to 1.4220 (20°C, 589 nm)
Residue after evaporation=<0.01 %
Appearance (Form)Clear liquid
Peroxides=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only