2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™
Thermo Scientific Chemicals

2-Methyltetrahydrofuran, 99+%, Extra Dry, stabilized, AcroSeal™

CAS: 96-47-9 | C5H10O | 86.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 396621000
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
106.65
Special offer
Online exclusive
Ends: 28-Dec-2026
125.00
Save 18.35
Each
Chemical Identifiers
CAS96-47-9
IUPAC Name2-methyloxolane
Molecular FormulaC5H10O
InChI KeyJWUJQDFVADABEY-UHFFFAOYNA-N
SMILESCC1CCCO1
View more
SpecificationsSpecification SheetSpecification Sheet
Peroxides=<50 ppm
Appearance (Form)Liquid
Water=<0.002 %
Appearance (Color)Clear colorless
GC>=99.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only