1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
Thermo Scientific Chemicals

1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 396651000
Price (USD)
620.65
Special offer
Online exclusive
Ends: 28-Dec-2026
730.00
Save 109.35
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
620.65
Special offer
Online exclusive
Ends: 28-Dec-2026
730.00
Save 109.35
Each
Chemical Identifiers
CAS22767-49-3
IUPAC Namesodium pentane-1-sulfonate
Molecular FormulaC5H11NaO3S
InChI KeyROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES[Na+].CCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV(0.5 M in water)
UVat 220 nm A: =<0.06
Titration after Ion exchange>=99.0 % (on dry substance)
UVat 230 nm A: =<0.04
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only