1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous
Thermo Scientific Chemicals

1-Hexanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous

CAS: 2832-45-3 | C6H13NaO3S | 188.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 396670250
Price (USD)
416.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
416.00
Each
Chemical Identifiers
CAS2832-45-3
IUPAC Namesodium hexane-1-sulfonate
Molecular FormulaC6H13NaO3S
InChI KeyQWSZRRAAFHGKCH-UHFFFAOYSA-M
SMILES[Na+].CCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline solid
UVat 210 nm A: =<0.1
UVat 230 nm A: =<0.04
Suitability(HPLC gradient) Passes test
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only