Methyl 4-(aminomethyl)benzoate hydrochloride, 97%
Methyl 4-(aminomethyl)benzoate hydrochloride, 97%
Methyl 4-(aminomethyl)benzoate hydrochloride, 97%
Thermo Scientific Chemicals

Methyl 4-(aminomethyl)benzoate hydrochloride, 97%

CAS: 6232-11-7 | C9H11NO2·ClH | 201.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 396780250
Price (USD)
206.65
Special offer
Online exclusive
Ends: 28-Dec-2026
243.00
Save 36.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
206.65
Special offer
Online exclusive
Ends: 28-Dec-2026
243.00
Save 36.35
Each
Chemical Identifiers
CAS6232-11-7
IUPAC Namemethyl 4-(aminomethyl)benzoate hydrochloride
Molecular FormulaC9H12ClNO2
InChI KeyGIZCKBSSWNIUMZ-UHFFFAOYSA-N
SMILESCl.COC(=O)C1=CC=C(CN)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Argentometric96.0 to 104.0 %
Appearance (Color)White
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only