D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals
D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals
D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

D-Serine methyleester hydrochloride, 98%, Thermo Scientific Chemicals

CAS: 5874-57-7 | Molecular Formula: C23H19Br2N3O3S | Molecular Weight: 577.29 g/mol
Have Questions?
Catalog NumberQuantityPackaging
39679025025 gGlass Bottle
Catalog number 396790250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS5874-57-7
IUPAC Name4-bromo-N-{1-[3-(4-bromophenyl)-4-oxo-3,4-dihydroquinazolin-2-yl]ethyl}-N-methylbenzene-1-sulfonamide
Molecular FormulaC23H19Br2N3O3S
InChI KeyLYRWWUCOZBTOPL-UHFFFAOYNA-N
SMILESCC(N(C)S(=O)(=O)C1=CC=C(Br)C=C1)C1=NC2=CC=CC=C2C(=O)N1C1=CC=C(Br)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Specific optical rotation-2.5° to -4.5° (20°C, 589 nm) (C=4, methanol)
Titration Argentometric>=97.5 %
Appearance (Color)White

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only