Benzyl alcohol, 98+%, Extra Dry, AcroSeal™
Benzyl alcohol, 98+%, Extra Dry, AcroSeal™
Benzyl alcohol, 98+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Benzyl alcohol, 98+%, Extra Dry, AcroSeal™

CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 396881000
Price (USD)
152.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (USD)
152.00
Each
Chemical Identifiers
CAS100-51-6
IUPAC Namephenylmethanol
Molecular FormulaC7H8O
InChI KeyWVDDGKGOMKODPV-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Residue after evaporation=<0.05 %
Appearance (Form)Clear liquid
GC>=98.0 %
Color scale=<20 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only