Di-n-butyl ether, 99+%, Extra Dry, AcroSeal™
Di-n-butyl ether, 99+%, Extra Dry, AcroSeal™
Di-n-butyl ether, 99+%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

Di-n-butyl ether, 99+%, Extra Dry, AcroSeal™

CAS: 142-96-1 | C8H18O | 130.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
Catalog number 396910025
Price (USD)
-
Quantity:
2.5 L
Request bulk or custom format
Chemical Identifiers
CAS142-96-1
IUPAC Name1-butoxybutane
Molecular FormulaC8H18O
InChI KeyDURPTKYDGMDSBL-UHFFFAOYSA-N
SMILESCCCCOCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.002 %
GC>=99.0 %
Infrared spectrumConforms
Water=<0.005 %
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only