Isopropanol, 99+%, specified according to the requirements of Ph.Eur.
Isopropanol, 99+%, specified according to the requirements of Ph.Eur.
Isopropanol, 99+%, specified according to the requirements of Ph.Eur.
Thermo Scientific Chemicals

Isopropanol, 99+%, specified according to the requirements of Ph.Eur.

CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
10 L
25 L
Catalog number 396980025
Price (USD)
92.00
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
92.00
Each
Chemical Identifiers
CAS67-63-0
IUPAC Namepropan-2-ol
Molecular FormulaC3H8O
InChI KeyKFZMGEQAYNKOFK-UHFFFAOYSA-N
SMILESCC(C)O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(25°C) 0.783 to 0.787
Identification DPasses test
Non-volatile matter=<20 ppm
Water=<0.5 %
UVat 290 nm A: =<0.02
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only