Sarcosine methyl ester, hydrochloride, 97%
Sarcosine methyl ester, hydrochloride, 97%
Sarcosine methyl ester, hydrochloride, 97%
Thermo Scientific Chemicals

Sarcosine methyl ester, hydrochloride, 97%

CAS: 13515-93-0 | C4H10ClNO2 | 139.58 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Glass bottle
Catalog number 397500500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS13515-93-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with HClO4>=96.0 %
Loss on drying=<1.0 % (1 g, 70°C) (to constant weight)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only