(tert-Butyldimethylsiloxy)acetaldehyde, 90%
(tert-Butyldimethylsiloxy)acetaldehyde, 90%
(tert-Butyldimethylsiloxy)acetaldehyde, 90%
Thermo Scientific Chemicals

(tert-Butyldimethylsiloxy)acetaldehyde, 90%

CAS: 102191-92-4 | C8H18O2Si | 174.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 397510050
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
268.65
Online Exclusive
298.00
Save 29.35 (10%)
Each
Chemical Identifiers
CAS102191-92-4
IUPAC Name2-[(tert-butyldimethylsilyl)oxy]acetaldehyde
Molecular FormulaC8H18O2Si
InChI KeyMEBFFOKESLAUSJ-UHFFFAOYSA-N
SMILESCC(C)(C)[Si](C)(C)OCC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=88 %
Appearance (Color)Clear colorless to straw yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only