What type of glass are the Glass Beads, 500 to 750um made out of?
The Glass Beads, 500 to 750µm are made of soda lime glass.
Search
View additional product information for Glass beads, 400-800 micron - FAQs (397640250, 397641000, 397645000)
1 product FAQs found