Glass beads, 400-800 micron - FAQs
View additional product information for Glass beads, 400-800 micron - FAQs (397640250, 397641000, 397645000)
1 product FAQs found
What type of glass are the Glass Beads, 500 to 750um made out of?
The Glass Beads, 500 to 750µm are made of soda lime glass.