4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
4-(Trifluoromethyl)benzenesulfonyl chloride, 98%
Thermo Scientific Chemicals

4-(Trifluoromethyl)benzenesulfonyl chloride, 98%

CAS: 2991-42-6 | C7H4ClF3O2S | 244.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 397760010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS2991-42-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only