2-Chloro-5-pyridineboronic acid, 97%
2-Chloro-5-pyridineboronic acid, 97%
2-Chloro-5-pyridineboronic acid, 97%
Thermo Scientific Chemicals

2-Chloro-5-pyridineboronic acid, 97%

CAS: 444120-91-6 | C5H5BClNO2 | 157.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 397770050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS444120-91-6
IUPAC Name(6-chloropyridin-3-yl)boronic acid
Molecular FormulaC5H5BClNO2
InChI KeyWPAPNCXMYWRTTL-UHFFFAOYSA-N
SMILESOB(O)C1=CC=C(Cl)N=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point161°C to 167°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only