3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%
3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%
3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%
Thermo Scientific Chemicals

3,3,3-Trifluoro-2-(trifluoromethyl)propionic acid, 97%

CAS: 564-10-3 | C4H2F6O2 | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 397860010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS564-10-3
IUPAC Name3,3,3-trifluoro-2-(trifluoromethyl)propanoic acid
Molecular FormulaC4H2F6O2
InChI KeyRAEAYTICAPHWJW-UHFFFAOYSA-N
SMILESOC(=O)C(C(F)(F)F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point49°C to 53°C
Titration with NaOH>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only