3-Bromo-4-pyridinecarboxaldehyde, 97%
3-Bromo-4-pyridinecarboxaldehyde, 97%
3-Bromo-4-pyridinecarboxaldehyde, 97%
Thermo Scientific Chemicals

3-Bromo-4-pyridinecarboxaldehyde, 97%

CAS: 70201-43-3 | C6H4BrNO | 186 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3980800101 gGlass bottle
Catalog number 398080010
Price (USD)
139.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
139.00
Each
Chemical Identifiers
CAS70201-43-3
IUPAC Name3-bromopyridine-4-carbaldehyde
Molecular FormulaC6H4BrNO
InChI KeyNOBDKWLIAQKADB-UHFFFAOYSA-N
SMILESBrC1=C(C=O)C=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Light yellow to light brown
Infrared spectrumConforms
NMR (1H-NMR)Consistent with structure
Melting point79°C to 83°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only