(R)-(+)-N-BOC-3-aminopyrrolidine, 97%
(R)-(+)-N-BOC-3-aminopyrrolidine, 97%
(R)-(+)-N-BOC-3-aminopyrrolidine, 97%
Thermo Scientific Chemicals

(R)-(+)-N-BOC-3-aminopyrrolidine, 97%

CAS: 147081-49-0 | C9H18N2O2 | 186.25 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 398270050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS147081-49-0
IUPAC Nametert-butyl 3-aminopyrrolidine-1-carboxylate
Molecular FormulaC9H18N2O2
InChI KeyCMIBWIAICVBURI-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCC(N)C1
さらに表示
Appearance (Color)Clear colourless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=96.0 %
Specific optical rotation+2° to +4° (20°C, 589 nm) (c=1 in CHCl3)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only