2,4-Dichloro-5-methylpyrimidine, 98%
2,4-Dichloro-5-methylpyrimidine, 98%
2,4-Dichloro-5-methylpyrimidine, 98%
Thermo Scientific Chemicals

2,4-Dichloro-5-methylpyrimidine, 98%

CAS: 1780-31-0 | C5H4Cl2N2 | 163.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 398290050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1780-31-0
IUPAC Name2,4-dichloro-5-methylpyrimidine
Molecular FormulaC5H4Cl2N2
InChI KeyDQXNTSXKIUZJJS-UHFFFAOYSA-N
SMILESCC1=CN=C(Cl)N=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)Light yellow
Appearance (Form)Low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only