5-Azabenzimidazole, 97%
5-Azabenzimidazole, 97%
5-Azabenzimidazole, 97%
Thermo Scientific Chemicals

5-Azabenzimidazole, 97%

CAS: 272-97-9 | C6H5N3 | 119.13 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3985000101 gGlass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 398500010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS272-97-9
IUPAC Name3H-imidazo[4,5-c]pyridine
Molecular FormulaC6H5N3
InChI KeyUBOOKRVGOBKDMM-UHFFFAOYSA-N
SMILESN1C=NC2=CC=NC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Beige
Appearance (Form)Powder
Infrared spectrumConforms
Melting point168°C to 172°C
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only