5-Bromoindole-2-carboxylic acid, 98%
5-Bromoindole-2-carboxylic acid, 98%
5-Bromoindole-2-carboxylic acid, 98%
Thermo Scientific Chemicals

5-Bromoindole-2-carboxylic acid, 98%

CAS: 7254-19-5 | C9H6BrNO2 | 240.05 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 398650050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7254-19-5
IUPAC Name5-bromo-1H-indole-2-carboxylic acid
Molecular FormulaC9H6BrNO2
InChI KeyYAULOOYNCJDPPU-UHFFFAOYSA-N
SMILESOC(=O)C1=CC2=CC(Br)=CC=C2N1
さらに表示
Appearance (Color)White to beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only