4-Iodophenylhydrazine, 95%
4-Iodophenylhydrazine, 95%
4-Iodophenylhydrazine, 95%
Thermo Scientific Chemicals

4-Iodophenylhydrazine, 95%

CAS: 13116-27-3 | C6H7IN2 | 234.04 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 399050050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS13116-27-3
IUPAC Name(4-iodophenyl)hydrazine
Molecular FormulaC6H7IN2
InChI KeyIQMLCMKMSBMMGR-UHFFFAOYSA-N
SMILESNNC1=CC=C(I)C=C1
さらに表示
Appearance (Color)White to orange to brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point99°C to 106°C
NMR>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only