Methyl 3-iodo-4-methoxybenzoate, 98%
Methyl 3-iodo-4-methoxybenzoate, 98%
Methyl 3-iodo-4-methoxybenzoate, 98%
Methyl 3-iodo-4-methoxybenzoate, 98%
Thermo Scientific Chemicals

Methyl 3-iodo-4-methoxybenzoate, 98%

CAS: 35387-93-0 | C9H9IO3 | 292.07 g/mol
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 399090250
Price (JPY)
-
Request A Quote
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS35387-93-0
IUPAC Namemethyl 3-iodo-4-methoxybenzoate
Molecular FormulaC9H9IO3
InChI KeyGHNGBFHLUOJHKP-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(OC)C(I)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to off-white
Melting point91°C to 95°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only