Ethyl 4,4,4-trifluoro-2-butynoate, 97%
Ethyl 4,4,4-trifluoro-2-butynoate, 97%
Ethyl 4,4,4-trifluoro-2-butynoate, 97%
Thermo Scientific Chemicals

Ethyl 4,4,4-trifluoro-2-butynoate, 97%

CAS: 79424-03-6 | C6H5F3O2 | 166.099 g/mol
Have Questions?
Catalog NumberQuantityPackaging
399325000500 mgGlass bottle
Catalog number 399325000
Price (USD)
-
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS79424-03-6
IUPAC Nameethyl 4,4,4-trifluorobut-2-ynoate
Molecular FormulaC6H5F3O2
InChI KeySFDRHPQGYUYYNX-UHFFFAOYSA-N
SMILESCCOC(=O)C#CC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Appearance (Form)Liquid
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only