BOC-L-alpha-cyclohexylglycine, 98%
BOC-L-alpha-cyclohexylglycine, 98%
BOC-L-alpha-cyclohexylglycine, 98%
Thermo Scientific Chemicals

BOC-L-alpha-cyclohexylglycine, 98%

CAS: 109183-71-3 | C13H23NO4 | 257.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 399350010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS109183-71-3
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}-2-cyclohexylacetic acid
Molecular FormulaC13H23NO4
InChI KeyQSUXZIPXYDQFCX-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)NC(C1CCCCC1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
NMRconsistent with structure
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only