N-BOC-3-Piperidinecarboxylic acid, 98%
N-BOC-3-Piperidinecarboxylic acid, 98%
N-BOC-3-Piperidinecarboxylic acid, 98%
Thermo Scientific Chemicals

N-BOC-3-Piperidinecarboxylic acid, 98%

CAS: 84358-12-3 | C11H19NO4 | 229.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 399580250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS84358-12-3
IUPAC Name1-[(tert-butoxy)carbonyl]piperidine-3-carboxylic acid
Molecular FormulaC11H19NO4
InChI KeyNXILIHONWRXHFA-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point159°C to 162°C
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only