6-Methoxy-2-naphthaleneboronic acid, 97%
6-Methoxy-2-naphthaleneboronic acid, 97%
6-Methoxy-2-naphthaleneboronic acid, 97%
Thermo Scientific Chemicals

6-Methoxy-2-naphthaleneboronic acid, 97%

CAS: 156641-98-4 | C11H11BO3 | 202.02 g/mol
製品番号(カタログ番号)数量
3998600505 g
製品番号(カタログ番号) 399860050
価格(JPY)
-
数量:
5 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS156641-98-4
IUPAC Name(6-methoxynaphthalen-2-yl)boronic acid
Molecular FormulaC11H11BO3
InChI KeyGZFAVYWCPSMLCM-UHFFFAOYSA-N
SMILESCOC1=CC2=CC=C(C=C2C=C1)B(O)O
さらに表示
NMR (1H-NMR)Consistent with structure
Appearance (Color)White to off-white to light brown
HPLC>=96.0 %
Appearance (Form)Powder or crystalline powder
Infrared spectrumAuthentic
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only