Cryopreservation Vials, Canes and Canisters
Actual product may vary
Cryopreservation Vials, Canes and Canisters
Cryopreservation Vials, Canes and Canisters
Thermo Scientific™
Cryopreservation Vials, Canes and Canisters
Maximize storage capacities in the laboratory by using these Cryopreservation Vials, Canes and Canisters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
4000198Cryo Vial1.2 Cardboard Boxes
4000200Cryo Vial2.0 Cardboard Boxes
4000206Cryo Vial4.0 Cardboard Boxes
4000208Cryo Vial5.0 Cardboard Boxes
4000211Cryo Cane6 Ampules
4000214Cryo Cane2 Ampules
4000216Cryo Cane9.2 and 10 mL Goblets
4000217Cryo Cane12.5 mm dia. Cryo Tubes
4000389Cryo Tube Color Coding CapsCryo Vials
Catalog number 4000198
Price (USD)
-
Type:
Cryo Vial
For Use With (Equipment):
1.2 Cardboard Boxes
Maximize storage capacities using Thermo Scientific™ Cryopreservation Vials, Canes and Canisters.
  • Warranty varies by country, please contact us for details
Specifications
DescriptionSelf-standing, conical interior bottom, silicone washer, case of 500
For Use With (Equipment)1.2 Cardboard Boxes
TypeCryo Vial
Capacity (English)4.23 cu. ft.
Capacity (Metric)1.2 mL
Unit SizeEach