4-Acetamidobenzaldehyde, 98%
4-Acetamidobenzaldehyde, 98%
4-Acetamidobenzaldehyde, 98%
Thermo Scientific Chemicals

4-Acetamidobenzaldehyde, 98%

CAS: 122-85-0 | C9H9NO2 | 163.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 400020100
Price (TWD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS122-85-0
IUPAC NameN-(4-formylphenyl)acetamide
Molecular FormulaC9H9NO2
InChI KeySKLUWKYNZNXSLX-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point154°C to 158°C
GC>=97.5 %
Appearance (Form)Crystalline powder
Appearance (Color)White to yellow
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only