Cryo Marker Pen Set
Thermo Scientific™

Cryo Marker Pen Set

Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColor
4000222Black
4000221Multi-color
Catalog number 4000222
Price (CLP)
-
Color:
Black

Label cryogenic vials to -200°C (-328°F) with the Thermo Scientific™ Cryo Marker Pen Set. The ink is fast-drying, nonsmearing and permanent.

  • Multicolor set is available; includes 4 fine-tip pens in black, blue, green and red
  • Warranty varies by country, please contact us for details
Specifications
ColorBlack
For Use With (Equipment)cryovials and boxes
Includes4 black pens
TypeCryo Marker Pen Set
Temperature Range-200°C, -328°F
Unit SizeBOX