Racks for the CryoMed™ Controlled-Rate Freezer
Actual product may vary
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Racks for the CryoMed™ Controlled-Rate Freezer
Thermo Scientific™
Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsProduct Type
400034010 Canisters (for cord blood, Model 4000610)Canister Rack
185089Adjustable, 10-positionCanister Rack
400029440 0.25/0.5 mL StrawsStraw Rack
4000303162 Canes, 5 to 6 Ampules/CaneCane Freezing Rack
400031048 Skin Packets (2 level)Skin Rack
400031172 Packets (3 level)Skin Rack
400031224 Packets (1 level)Skin Rack
400070077 CanesCane Freezing Rack
400070176 1.2 mL/2 mL AmpulesVial Rack (Small)
40007025 x 1.2/2 mL Cryo Racks (Small)Vial Rack Holder
4000703161 1.2 mL/2 mL AmpulesVial Rack (Large)
40007045 x 1.2/2 mL Cryo Racks (Large)Vial Rack Holder
400070576 4 mL/5 mL AmpulesVial Rack (Small)
40007063 x 4/5 mL Cryo Racks (Small)Vial Rack Holder
4000707161 4 mL/5 mL AmpulesVial Rack (Large)
40007083 x 4/5 mL Cryo Racks (Large)Vial Rack Holder
Catalog number 4000340
Price (MXN)
-
Holds:
10 Canisters (for cord blood, Model 4000610)
Product Type:
Canister Rack
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
  • Warranty varies by country, please contact us for details
Specifications
Description10 Canisters (for cord blood, Model 4000610)
For Use With (Equipment)All CryoMed controlled-rate freezers
Holds10 Canisters (for cord blood, Model 4000610)
ColorSilver
Product TypeCanister Rack
Unit SizeEach