CryoMed™ Thermocouple Sensors
Actual product may vary
CryoMed™ Thermocouple Sensors
CryoMed™ Thermocouple Sensors
Thermo Scientific™
CryoMed™ Thermocouple Sensors
Sample thermocouple sensors
Have Questions?
카탈로그 번호유형
4000393Ribbon Type
카탈로그 번호 4000393
제품 가격(KRW)
-
유형:
Ribbon Type
Sample thermocouple sensors
Thermocouple sensors for cryo tubes, sheath straw and vial samples.
  • Include male plug
For controlled-rate freezing applications
사양
보유Cryo Tube, Sheath Straw, Vial
유형Ribbon Type
플러그 유형Male
Unit SizeEach