RapidHIT™ ID ACE GlobalFiler™ Express Evaluation Kit
Applied Biosystems™

RapidHIT™ ID ACE GlobalFiler™ Express Evaluation Kit

The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Evaluation Kit is used with the Applied Biosystems RapidHIT ID System toRead more
Have Questions?
Catalog NumberQuantity
400114RH10 cartridges kit
Catalog number 400114RH
Price (MXN)
-
Quantity:
10 cartridges kit

The Applied Biosystems RapidHIT ID ACE GlobalFiler Express Evaluation Kit is used with the Applied Biosystems RapidHIT ID System to generate lab-quality forensic DNA profiles primarily from reference samples collected from known individuals. These DNA profiles can be compared to DNA profiles obtained from crime scene evidence.

This kit contains 10 ACE GlobalFiler Express Sample Cartridges, each of which is an individually-wrapped, single-use, disposable cartridge that contains reagents for developing a forensic DNA profile from a single sample in 90 minutes.

To run crime scene samples on the Applied Biosystems RapidHIT ID System, see our Applied Biosystems RapidINTEL Sample Cartridge Kit.

Intended use:
• Reference samples and high quantity crime scene samples

Required for use:
• Applied Biosystems RapidHIT ID System (Cat. No. A41810)
• Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
• Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)

Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID System
No. of Samples10 Samples
Quantity10 cartridges kit
Sample TypeBuccal swabs, High DNA content crime scene samples
Shelf Life12 Months
FormatKit
Unit SizeEach