Applied Biosystems™

R1 (5% PITC in N-Heptane)

Catalog number:  400208

 Related applications: Protein Sequencing & Peptide Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

5% PITC (phenylisothiocynate) in heptane is used for N-terminal protein sequencing applications. PITC is a derivatization reagent used to tag amino acids in the protein sequencer and amino acid analyzer. An optimal PITC concentration is required to ensure efficient amino acid coupling with minimal background contribution from excess reagent. This reagent is used in conjunction with other protein sequencing reagents.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Form: Liquid
Product Size: 40 mL
Reagent Type: N-terminal Sequencing Reagent
Shipping Condition: Room Temperature

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols