Product Image
Applied Biosystems™
Bulk CPG (Controlled Pore Glass), 500 Angstrom Pore Size, dC(Bz)
Have Questions?
Catalog NumberQuantity
4003951 g
Catalog number 400395
Price (USD)
1,400.00
1 g
Quantity:
1 g
Price (USD)
1,400.00
1 g
Bulk dC(Bz) Controlled Pore Glass (CPG) for users to pack their own columns for synthesis of oligonucleotides <50 bases in length.
• Proprietary design and manufacturing methods ensure consistency and high quality
• The 500 angstrom pore size of the support allows synthesis of oligonucleotides of up to 50 bases
• Save time and money
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeOligonucleotide Synthesis Column
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
MatrixControlled Pore Glass
Pore Size500 Å
Product TypeOligonucleotide Synthesis Columns
Quantity1 g
Sample TypeOligos
Shipping ConditionRoom Temperature
TargetNucleic Acids
FormatBottle
Unit Size1 g
Contents & Storage
Store bulk support in a dry environment at room temperature.