DNA Synthesis Column, Empty, 10.0 µmol crimp style

Catalog number: 400408

 Related applications: Nucleic Acid Labeling & Oligo Synthesis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This set of 10 empty columns allows users to pack their own DNA synthesis columns.

For Research Use Only. Not for use in diagnostics procedures.

Specifications

Column Type: Oligonucleotide Synthesis Column
For Use With (Equipment): 3900 DNA Synthesizer
Format: Column
Product Line: Applied Biosystems™
Product Size: 10 units
Sample Type (Specific): Oligos
Shipping Condition: Room Temperature

Contents & storage

Store at room temperature.

Documents

Manuals & protocols