Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Alizarin Red S, pure, certified
Thermo Scientific Chemicals

Alizarin Red S, pure, certified

CAS: 130-22-3 | C14H7NaO7S | 342.253 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 400481000
Price (USD)
191.00
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
191.00
Each
Chemical Identifiers
CAS130-22-3
IUPAC Namesodium 3,4-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Molecular FormulaC14H7NaO7S
InChI KeyHFVAFDPGUJEFBQ-UHFFFAOYSA-M
SMILES[Na+].OC1=C(O)C2=C(C=C1S([O-])(=O)=O)C(=O)C1=CC=CC=C1C2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Visual transition intervalfrom pH 3.5 (yellow) to pH 6.5 (pink)
Appearance (Color)Yellow to orange to brown
Infrared spectrumConforms
UV Lambda max554 to 558 nm (in 0.1 M NaOH)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only