4'-Aminoacetanilide, 95%
4'-Aminoacetanilide, 95%
4'-Aminoacetanilide, 95%
Thermo Scientific Chemicals

4'-Aminoacetanilide, 95%

CAS: 122-80-5 | C8H10N2O | 150.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 400601000
Price (USD)
158.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
158.00
Each
Chemical Identifiers
CAS122-80-5
IUPAC NameN-(4-aminophenyl)acetamide
Molecular FormulaC8H10N2O
InChI KeyCHMBIJAOCISYEW-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pink to brown
Appearance (Form)Crystalline powder
Melting point159°C to 165°C
Infrared spectrumConforms
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only