4-(2-Aminoethyl)morpholine, 98+%
4-(2-Aminoethyl)morpholine, 98+%
4-(2-Aminoethyl)morpholine, 98+%
4-(2-Aminoethyl)morpholine, 98+%
Thermo Scientific Chemicals

4-(2-Aminoethyl)morpholine, 98+%

CAS: 2038-03-1 | C6H14N2O | 130.19 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 400751000
価格(JPY)
-
見積もりを依頼する
数量:
100 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS2038-03-1
IUPAC Name2-(morpholin-4-yl)ethan-1-amine
Molecular FormulaC6H14N2O
InChI KeyRWIVICVCHVMHMU-UHFFFAOYSA-N
SMILESNCCN1CCOCC1
さらに表示
Appearance (Color)Clear colorless to white to yellow
Appearance (Form)Liquid or solid
Refractive index1.4740 to 1.4780 (20°C, 589 nm)
GC>=98.0 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only