4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent
Thermo Scientific Chemicals

4-Amino-3-hydroxy-1-naphthalenesulfonic acid, 90+%, ACS reagent

CAS: 116-63-2 | C10H9NO4S | 239.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 400850250
Price (EUR)
94,65
Online exclusive
105,00
Save 10,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
94,65
Online exclusive
105,00
Save 10,35 (10%)
Each
Chemical Identifiers
CAS116-63-2
IUPAC Name4-amino-3-hydroxynaphthalene-1-sulfonic acid
Molecular FormulaC10H9NO4S
InChI KeyRXCMFQDTWCCLBL-UHFFFAOYSA-N
SMILESNC1=C2C=CC=CC2=C(C=C1O)S(O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown-pink to light purple
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=90.0 %
Solubility(in sodium carbonate) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only