2-Amino-5-nitrophenol, 95%
2-Amino-5-nitrophenol, 95%
2-Amino-5-nitrophenol, 95%
Thermo Scientific Chemicals

2-Amino-5-nitrophenol, 95%

CAS: 121-88-0 | C6H6N2O3 | 154.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 400900010
Price (EUR)
737,00
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
737,00
Each
Chemical Identifiers
CAS121-88-0
IUPAC Name2-amino-5-nitrophenol
Molecular FormulaC6H6N2O3
InChI KeyDOPJTDJKZNWLRB-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1O)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Rust brown to brown
Appearance (Form)Crystalline powder
Titration with HClO4>=94 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only